1-BOC-4-CBZ-2-(Hydroxymethyl)piperazine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-O-Benzyl 1-O-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate, 4-Benzyl 1-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate

OR914186-1G OR914186-250MG
APOSOR914186-1GEA 763 EUR
APOSOR914186-1G APOSOR914186-250MG
1-BOC-4-CBZ-2-(Hydroxymethyl)piperazine 95%
1-BOC-4-CBZ-2-(Hydroxymethyl)piperazine
Formel: C₁₈H₂₆N₂O₅
Molecular Weight: 350,409 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17676397
CAS-Nummer: 557056-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR