N-(2-Hydroxyethyl) 4-methyl-2-nitroaniline 98%

Lieferant: Apollo Scientific

OR914198-5G
APOSOR914198-5GEA 63.8 EUR
APOSOR914198-5G
N-(2-Hydroxyethyl) 4-methyl-2-nitroaniline 98%
N-(2-Hydroxyethyl) 4-methyl-2-nitroaniline
Formel: C₉H₁₂N₂O₃
Molecular Weight: 196,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274758
CAS-Nummer: 100418-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR