3-Bromo-5-hydroxy-2-picoline 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-methylpyridin-3-ol

OR914335-1G OR914335-250MG
APOSOR914335-1GEA 116 EUR
APOSOR914335-1G APOSOR914335-250MG
3-Bromo-5-hydroxy-2-picoline 98%
3-Bromo-5-hydroxy-2-picoline
Formel: C₆H₆BrNO
Molecular Weight: 188,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839298
CAS-Nummer: 186593-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR