1-(2-Methylpropoxy)-4-nitrobenzene 98%

Lieferant: Apollo Scientific

OR914336-1G OR914336-5G
APOSOR914336-1GEA 219 EUR
APOSOR914336-1G APOSOR914336-5G
1-(2-Methylpropoxy)-4-nitrobenzene 98%
1-(2-Methylpropoxy)-4-nitrobenzene
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21985346
CAS-Nummer: 57027-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR