1-(Cyclopropylmethoxy)-4-nitrobenzene 97%

Lieferant: Apollo Scientific

OR914356-1G OR914356-5G
APOSOR914356-1GEA 290 EUR
APOSOR914356-1G APOSOR914356-5G
1-(Cyclopropylmethoxy)-4-nitrobenzene 97%
1-(Cyclopropylmethoxy)-4-nitrobenzene
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09701724
CAS-Nummer: 85002-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR