ethyl 3-oxopyrrolidine-1-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-oxopyrrolidine-1-carboxylate

OR914467-1G OR914467-5G
APOSOR914467-1GEA 31.9 EUR
APOSOR914467-1G APOSOR914467-5G
ethyl 3-oxopyrrolidine-1-carboxylate 95%
ethyl 3-oxopyrrolidine-1-carboxylate
Formel: C₇H₁₁NO₃
Molecular Weight: 157,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093840
CAS-Nummer: 14891-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR