1-Bromo-2-methyl-4-(methylsulphonyl)benzene 96%

Lieferant: Apollo Scientific
Achtung

OR914495-1G OR914495-5G
APOSOR914495-1GEA 132 EUR
APOSOR914495-1G APOSOR914495-5G
1-Bromo-2-methyl-4-(methylsulphonyl)benzene 96%
1-Bromo-2-methyl-4-(methylsulphonyl)benzene
Formel: C₈H₉BrO₂S
Molecular Weight: 249,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22549330
CAS-Nummer: 99769-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR