4-Bromo-3,6-dihydro-2H-pyran 95%

Lieferant: Apollo Scientific

OR914531-10G OR914531-1G OR914531-5G
APOSOR914531-10GEA 964 EUR
APOSOR914531-10G APOSOR914531-1G APOSOR914531-5G
4-Bromo-3,6-dihydro-2H-pyran 95%
4-Bromo-3,6-dihydro-2H-pyran
Formel: C₅H₇BrO
Molecular Weight: 163.012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699698
CAS-Nummer: 24265-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR