2-Methoxy-3-methylnaphthalene 98%

Lieferant: Apollo Scientific

OR914701-1G OR914701-5G
APOSOR914701-1GEA 219 EUR
APOSOR914701-1G APOSOR914701-5G
2-Methoxy-3-methylnaphthalene 98%
2-Methoxy-3-methylnaphthalene
Formel: C₁₂H₁₂O
Molecular Weight: 172,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17012351
CAS-Nummer: 61873-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR