1-[(Methylsulphonyl)methyl]-4-nitrobenzene 98%

Lieferant: Apollo Scientific

OR915066-1G OR915066-25G OR915066-5G
APOSOR915066-1GEA 69.6 EUR
APOSOR915066-1G APOSOR915066-25G APOSOR915066-5G
1-[(Methylsulphonyl)methyl]-4-nitrobenzene 98%
1-[(Methylsulphonyl)methyl]-4-nitrobenzene
Formel: C₈H₉NO₄S
Schmelzpunkt: 170…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD27926301
CAS-Nummer: 61081-34-3
UN: 3272
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR