3-[(1H-Imidazol-1-yl)methyl]benzaldehyde 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(3-Formylbenzyl)-1H-imidazole

OR9153-1G OR9153-250MG
APOSOR9153-1GEA 198 EUR
APOSOR9153-1G APOSOR9153-250MG
3-[(1H-Imidazol-1-yl)methyl]benzaldehyde 97%
3-[(1H-Imidazol-1-yl)methyl]benzaldehyde
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186.21 g/mol
Siedepunkt: 185 °C (0.1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08271917
CAS-Nummer: 102432-05-3
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR