4-[(1H-Imidazol-1-yl)methyl]benzaldehyde 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-[(4-Formylphenyl)methyl]-1H-imidazole, 1-(4-Formylbenzyl)-1H-imidazole
OR9154-1G
OR9154-5G
APOSOR9154-1GEA
343
EUR
APOSOR9154-1G
APOSOR9154-5G
4-[(1H-Imidazol-1-yl)methyl]benzaldehyde 97%
4-[(1H-Imidazol-1-yl)methyl]benzaldehyde
Formel:
C₁₁H₁₀N₂O Molecular Weight: 186.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08271920 CAS-Nummer: 102432-03-1 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...