Furan-3-sulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Furan-3-sulphonyl chloride

OR9160-250MG OR9160-1G
APOSOR9160-250MGEA 234 EUR
APOSOR9160-250MG APOSOR9160-1G
Furan-3-sulphonyl chloride 97%
Furan-3-sulphonyl chloride
Formel: C₄H₃ClO₃S
Molecular Weight: 166,583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08271875
CAS-Nummer: 52665-49-3
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR