2-Chlorothiophene-3-carboxaldehyde 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chlorothiophene-3-carbaldehyde

OR916232-1G OR916232-5G
APOSOR916232-1GEA 97.3 EUR
APOSOR916232-1G APOSOR916232-5G
2-Chlorothiophene-3-carboxaldehyde 98%
2-Chlorothiophene-3-carboxaldehyde
Formel: C₅H₃ClOS
Molecular Weight: 146.595 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08694817
CAS-Nummer: 14345-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR