4-(Pyridin-2-yl)phenol 98%

Lieferant: Apollo Scientific

OR916258-5G OR916258-25G
APOSOR916258-5GEA 56.7 EUR
APOSOR916258-5G APOSOR916258-25G
4-(Pyridin-2-yl)phenol 98%
4-(Pyridin-2-yl)phenol
Formel: C₁₁H₉NO
Molecular Weight: 171.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464189
CAS-Nummer: 51035-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR