5-Methyl-1H-pyrrole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 5-Methylpyrrole-2-carbaldehyde

OR916381-1G OR916381-250MG OR916381-5G
APOSOR916381-1GEA 202 EUR
APOSOR916381-1G APOSOR916381-250MG APOSOR916381-5G
5-Methyl-1H-pyrrole-2-carbaldehyde 95%
5-Methyl-1H-pyrrole-2-carbaldehyde
Formel: C₆H₇NO
Molecular Weight: 109.126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08236773
CAS-Nummer: 1192-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR