7-Chloro-D-tryptophan 97%

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-3-(7-chloro-1H-indol-3-yl)propanoic acid, H-D-Trp(7-Cl)-OH

OR916429-250MG
APOSOR916429-250MGEA 331 EUR
APOSOR916429-250MG
7-Chloro-D-tryptophan 97%
7-Chloro-D-tryptophan
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264323
CAS-Nummer: 75102-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR