5-Methyl-1-(3-pyridinyl)-1H-pyrrole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR916541-1G OR916541-250MG
APOSOR916541-1GEA 400 EUR
APOSOR916541-1G APOSOR916541-250MG
5-Methyl-1-(3-pyridinyl)-1H-pyrrole-2-carbaldehyde 95%
5-Methyl-1-(3-pyridinyl)-1H-pyrrole-2-carbaldehyde
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09674953
CAS-Nummer: 931985-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR