2-Ethylpropane-1,3-diol 95%
Lieferant: Apollo Scientific
OR916592-1G
OR916592-5G
APOSOR916592-1GEA
226
EUR
APOSOR916592-1G
APOSOR916592-5G
2-Ethylpropane-1,3-diol 95%
2-Ethylpropane-1,3-diol
Formel:
C₅H₁₂O₂ Molecular Weight: 104.148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09909398 CAS-Nummer: 2612-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...