1-(6-Methylpyridin-2-yl)ethanamine 95%

Lieferant: Apollo Scientific

OR916658-1G OR916658-250MG
APOSOR916658-1GEA 864 EUR
APOSOR916658-1G APOSOR916658-250MG
1-(6-Methylpyridin-2-yl)ethanamine 95%
1-(6-Methylpyridin-2-yl)ethanamine
Formel: C₈H₁₂N₂
Molecular Weight: 136,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658491
CAS-Nummer: 58088-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR