3-(4,5-Dihydro-4,4-dimethyloxazol-2-yl)benzenamine 95%

Lieferant: Apollo Scientific

OR916913-1G OR916913-250MG
APOSOR916913-1GEA 734 EUR
APOSOR916913-1G APOSOR916913-250MG
3-(4,5-Dihydro-4,4-dimethyloxazol-2-yl)benzenamine 95%
3-(4,5-Dihydro-4,4-dimethyloxazol-2-yl)benzenamine
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909379
CAS-Nummer: 913731-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR