(4-Methoxy-phenyl)-oxo-acetonitrile 95%

Lieferant: Apollo Scientific

OR916987-1G OR916987-5G
APOSOR916987-1GEA 219 EUR
APOSOR916987-1G APOSOR916987-5G
(4-Methoxy-phenyl)-oxo-acetonitrile 95%
(4-Methoxy-phenyl)-oxo-acetonitrile
Formel: C₉H₇NO₂
Molecular Weight: 161,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658462
CAS-Nummer: 14271-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR