{4-[(1H-Imidazol-1-yl)methyl]phenyl}methanol 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-[4-(Hydroxymethyl)benzyl]-1H-imidazole, 4-[(1H-Imidazol-1-yl)methyl]benzyl alcohol

OR9170-1G OR9170-250MG
APOSOR9170-1GEA 321 EUR
APOSOR9170-1G APOSOR9170-250MG
{4-[(1H-Imidazol-1-yl)methyl]phenyl}methanol 97%
{4-[(1H-Imidazol-1-yl)methyl]phenyl}methanol
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188.226 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271921
CAS-Nummer: 103573-92-8
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR