Tetraacid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane, 3,3'-((2,2-Bis((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropanoic acid, 3-[3-(2-carboxyethoxy)-2,2-bis(2-carboxyethoxymethyl)propoxy]propanoic acid
OR917035-1G
OR917035-5G
OR917035-250MG
APOSOR917035-1GEA
328
EUR
APOSOR917035-1G
APOSOR917035-5G
APOSOR917035-250MG
Tetraacid
Tetraacid
Formel:
C₁₇H₂₈O₁₂ Molecular Weight: 424,397 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669605 CAS-Nummer: 35638-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...