4-(4-Aminophenoxy)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Pyridin-4-yloxy)aniline

OR917039-1G OR917039-250MG
APOSOR917039-1GEA 238 EUR
APOSOR917039-1G APOSOR917039-250MG
4-(4-Aminophenoxy)pyridine 95%
4-(4-Aminophenoxy)pyridine
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06825431
CAS-Nummer: 102877-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR