6-Methyl-1,3-benzodioxole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
OR917166-1G
OR917166-250MG
APOSOR917166-1GEA
400
EUR
APOSOR917166-1G
APOSOR917166-250MG
6-Methyl-1,3-benzodioxole-5-carbaldehyde 95%
6-Methyl-1,3-benzodioxole-5-carbaldehyde
Formel:
C₉H₈O₃ Molecular Weight: 164,158 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04041885 CAS-Nummer: 58343-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...