1H-Indole-6-acetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(1H-Indol-6-yl)acetic acid

OR917177-1G OR917177-250MG
APOSOR917177-1GEA 95.7 EUR
APOSOR917177-1G APOSOR917177-250MG
1H-Indole-6-acetic acid 98%
1H-Indole-6-acetic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09837606
CAS-Nummer: 39689-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR