(R)-tert-Butyl (2-oxoazepan-3-yl)carbamate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR917182-1G
OR917182-250MG
APOSOR917182-1GEA
142
EUR
APOSOR917182-1G
APOSOR917182-250MG
(R)-tert-Butyl (2-oxoazepan-3-yl)carbamate 95%
(R)-tert-Butyl (2-oxoazepan-3-yl)carbamate
Formel:
C₁₁H₂₀N₂O₃ Molecular Weight: 228,288 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03788637 CAS-Nummer: 106691-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...