4-(4,4-Dimethyl-4,5-dihydro-1,3-oxazol-2-yl)aniline 95%

Lieferant: Apollo Scientific

OR917191-1G OR917191-250MG
APOSOR917191-1GEA 734 EUR
APOSOR917191-1G APOSOR917191-250MG
4-(4,4-Dimethyl-4,5-dihydro-1,3-oxazol-2-yl)aniline 95%
4-(4,4-Dimethyl-4,5-dihydro-1,3-oxazol-2-yl)aniline
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02677755
CAS-Nummer: 69876-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR