3-[(1H-1,2,4-Triazol-1-yl)methyl]benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: {3-[(1H-1,2,4-Triazol-1-yl)methyl]phenyl}methanol

OR9173-1G OR9173-250MG
APOSOR9173-1GEA 183 EUR
APOSOR9173-1G APOSOR9173-250MG
3-[(1H-1,2,4-Triazol-1-yl)methyl]benzyl alcohol 95%
3-[(1H-1,2,4-Triazol-1-yl)methyl]benzyl alcohol
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189,214 g/mol
Schmelzpunkt: 56…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271929
CAS-Nummer: 871825-54-6
UN: 1325
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR