2,5-Dichloroquinoline 95%
Lieferant: Apollo Scientific
OR917331-1G
APOSOR917331-1GEA
58
EUR
APOSOR917331-1G
2,5-Dichloroquinoline 95%
2,5-Dichloroquinoline
Formel:
C₉H₅Cl₂N Molecular Weight: 198,049 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00832074 CAS-Nummer: 59412-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...