6-Chloro-4-nitro-1H-indazole 95%

Lieferant: Apollo Scientific

OR917449-1G OR917449-250MG
APOSOR917449-1GEA 127 EUR
APOSOR917449-1G APOSOR917449-250MG
6-Chloro-4-nitro-1H-indazole 95%
6-Chloro-4-nitro-1H-indazole
Formel: C₇H₄ClN₃O₂
Molecular Weight: 197,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781380
CAS-Nummer: 885519-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR