4-(Dimethylamino)phenol 97%
Lieferant: Apollo Scientific
OR917462-25G
OR917462-5G
APOSOR917462-25GEA
227
EUR
APOSOR917462-25G
APOSOR917462-5G
4-(Dimethylamino)phenol 97%
4-(Dimethylamino)phenol
Formel:
C₈H₁₁NO Molecular Weight: 137,179 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD01707548 CAS-Nummer: 619-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...