Methyl 3-(cyclopropylamino)propanoate 95%

Lieferant: Apollo Scientific

OR917489-1G OR917489-250MG
APOSOR917489-1GEA 609 EUR
APOSOR917489-1G APOSOR917489-250MG
Methyl 3-(cyclopropylamino)propanoate 95%
Methyl 3-(cyclopropylamino)propanoate
Formel: C₇H₁₃NO₂
Molecular Weight: 143,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11651272
CAS-Nummer: 77497-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR