(S)-2-(Methylamino)-3-phenylpropan-1-ol 95%
Lieferant: Apollo Scientific
OR917507-1G
APOSOR917507-1GEA
163
EUR
APOSOR917507-1G
(S)-2-(Methylamino)-3-phenylpropan-1-ol 95%
(S)-2-(Methylamino)-3-phenylpropan-1-ol
Formel:
C₁₀H₁₅NO Molecular Weight: 165,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18431682 CAS-Nummer: 84773-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...