Methyl 3-cyano-4-hydroxybenzoate 95%

Lieferant: Apollo Scientific

OR917547-1G OR917547-250MG
APOSOR917547-1GEA 243 EUR
APOSOR917547-1G APOSOR917547-250MG
Methyl 3-cyano-4-hydroxybenzoate 95%
Methyl 3-cyano-4-hydroxybenzoate
Formel: C₉H₇NO₃
Molecular Weight: 177,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13248596
CAS-Nummer: 156001-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR