7-Chloro-3,4-dihydro-2H-benzo[b]oxepin-5-one 95%

Lieferant: Apollo Scientific

OR917617-1G OR917617-250MG
APOSOR917617-1GEA 647 EUR
APOSOR917617-1G APOSOR917617-250MG
7-Chloro-3,4-dihydro-2H-benzo[b]oxepin-5-one 95%
7-Chloro-3,4-dihydro-2H-benzo[b]oxepin-5-one
Formel: C₁₀H₉ClO₂
Molecular Weight: 196,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01648758
CAS-Nummer: 55579-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR