2-(4-Bromophenyl)furan 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9177-1G
OR9177-250MG
APOSOR9177-1GEA
230
EUR
APOSOR9177-1G
APOSOR9177-250MG
2-(4-Bromophenyl)furan 97%
2-(4-Bromophenyl)furan
Formel:
C₁₀H₇BrO Molecular Weight: 223,066 g/mol Schmelzpunkt: 83…84,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07775614 CAS-Nummer: 14297-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...