3-Methyl-5-p-tolyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR917708-1G OR917708-250MG
APOSOR917708-1GEA 435 EUR
APOSOR917708-1G APOSOR917708-250MG
3-Methyl-5-p-tolyl-1H-pyrazole 95%
3-Methyl-5-p-tolyl-1H-pyrazole
Formel: C₁₁H₁₂N₂
Molecular Weight: 172.226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08446306
CAS-Nummer: 90861-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR