3,5-Dichlorophenethyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: 2-(3,5-Dichlorophenyl)ethanol

OR917823-10G OR917823-5G
APOSOR917823-10GEA 140 EUR
APOSOR917823-10G APOSOR917823-5G
3,5-Dichlorophenethyl alcohol 97%
3,5-Dichlorophenethyl alcohol
Formel: C₈H₈Cl₂O
Molecular Weight: 191,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06201039
CAS-Nummer: 93427-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR