trans-4-(Acetamidomethyl)cyclohexanecarboxylic acid 95%

Lieferant: Apollo Scientific

OR917890-1G OR917890-5G
APOSOR917890-1GEA 221 EUR
APOSOR917890-1G APOSOR917890-5G
trans-4-(Acetamidomethyl)cyclohexanecarboxylic acid 95%
trans-4-(Acetamidomethyl)cyclohexanecarboxylic acid
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00626152
CAS-Nummer: 20704-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR