4-Amino-2-chloropyridin-3-ol 95+%

Lieferant: Apollo Scientific
Achtung

OR917900-100MG OR917900-250MG
APOSOR917900-100MGEA 45.1 EUR
APOSOR917900-100MG APOSOR917900-250MG
4-Amino-2-chloropyridin-3-ol 95+%
4-Amino-2-chloropyridin-3-ol
Formel: C₅H₅ClN₂O
Molecular Weight: 144.559 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16606120
CAS-Nummer: 1227508-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR