5-Bromo-2-hydroxy-3-methylbenzonitrile 95+%

Lieferant: Apollo Scientific
Achtung

OR917916-1G OR917916-250MG OR917916-5G
APOSOR917916-1GEA 109 EUR
APOSOR917916-1G APOSOR917916-250MG APOSOR917916-5G
5-Bromo-2-hydroxy-3-methylbenzonitrile 95+%
5-Bromo-2-hydroxy-3-methylbenzonitrile
Formel: C₈H₆BrNO
Molecular Weight: 212.043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457442
CAS-Nummer: 913191-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR