4-Oxo-1-phenyl-cyclohexanecarboxylic acid 97%
Lieferant: Apollo Scientific
OR917952-1G
OR917952-250MG
APOSOR917952-1GEA
654
EUR
APOSOR917952-1G
APOSOR917952-250MG
4-Oxo-1-phenyl-cyclohexanecarboxylic acid 97%
4-Oxo-1-phenyl-cyclohexanecarboxylic acid
Formel:
C₁₃H₁₄O₃ Molecular Weight: 218.248 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06208311 CAS-Nummer: 75945-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...