N-Methyl-1H-1,3-benzodiazol-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-1H-benzo[d]imidazol-2-amine

OR917990-1G OR917990-250MG
APOSOR917990-1GEA 277 EUR
APOSOR917990-1G APOSOR917990-250MG
N-Methyl-1H-1,3-benzodiazol-2-amine 95%
N-Methyl-1H-1,3-benzodiazol-2-amine
Formel: C₈H₉N₃
Molecular Weight: 147.177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00524803
CAS-Nummer: 17228-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR