1-Bromocyclobutane-1-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
1-Bromocyclobutanecarboxylic acid
OR918097-10G
OR918097-25G
APOSOR918097-10GEA
0
EUR
APOSOR918097-10G
APOSOR918097-25G
1-Bromocyclobutane-1-carboxylic acid
1-Bromocyclobutane-1-carboxylic acid
Formel:
C₅H₇BrO₂ Molecular Weight: 179,012 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08861855 CAS-Nummer: 32122-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...