1-Methyl-4-nitronaphthalene 95%

Lieferant: Apollo Scientific

OR918208-1G OR918208-5G
APOSOR918208-1GEA 133 EUR
APOSOR918208-1G APOSOR918208-5G
1-Methyl-4-nitronaphthalene 95%
1-Methyl-4-nitronaphthalene
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00971804
CAS-Nummer: 880-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR