4-Amino-2-methylbenzonitrile 95%

Lieferant: Apollo Scientific
Achtung

OR918260-1G OR918260-250MG OR918260-5G
APOSOR918260-1GEA 58 EUR
APOSOR918260-1G APOSOR918260-250MG APOSOR918260-5G
4-Amino-2-methylbenzonitrile 95%
4-Amino-2-methylbenzonitrile
Formel: C₈H₈N₂
Molecular Weight: 132,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260793
CAS-Nummer: 72115-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR