Methylsuccinic anhydride 98%

Lieferant: Apollo Scientific

OR918278-100G OR918278-25G
APOSOR918278-100GEA 0 EUR
APOSOR918278-100G APOSOR918278-25G
Methylsuccinic anhydride 98%
Methylsuccinic anhydride
Formel: C₅H₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00005527
CAS-Nummer: 4100-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR