Methyl 2-chloro-4-cyanobenzoate 95%

Lieferant: Apollo Scientific

OR918297-1G OR918297-5G
APOSOR918297-1GEA 198 EUR
APOSOR918297-1G APOSOR918297-5G
Methyl 2-chloro-4-cyanobenzoate 95%
Methyl 2-chloro-4-cyanobenzoate
Formel: C₉H₆ClNO₂
Molecular Weight: 195,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14635863
CAS-Nummer: 98592-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR